Learn More
ACROBiosystems ACROBiosystems Canine IL-3 R alpha / CD123 Protein, His Tag
Catalog No. 502106182
Encompass_Preferred
Recombinant Protein;100UG;Canine IL-3 R alpha, His Tag (ILA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Asp 315 (Accession # XP_038305195.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6182 Supplier ACROBiosystems Supplier No. ILAC52H3100UG
Recombinant Protein;100UG;Canine IL-3 R alpha, His Tag (ILA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Asp 315 (Accession # XP_038305195.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.