Learn More
Kingfisher Biotech Inc Canine IL-36RA Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1997D100
Encompass
The Canine IL-36 Receptor Antagonist yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IL-36 Receptor Antagonist applications are for cell culture, ELISA standard, and Western Blot Control. Canine IL-36 Receptor Antagonist yeast-derived recombinant protein can be purchased in multiple sizes. Canine IL-36 Receptor Antagonist Specifications: (Molecular Weight: 16.9 kDa) (Amino Acid Sequence: VLSGALCFRM KDAALKVLYL HDNQLLAGGL HAGKVIKGEE ISVVPNRSLD AKLSPVILGV QGGSQCLSCG TGQEPTLKLE PVNIMELYLG AKESKSFTFY RRDTGLTSSF ESAAYPGWFL CTVPEADQPL RLTQLSEDAS WDNPITDFYF QQCD (154)) (Gene ID: 611869). For research use only. Made in the USA
Catalog No. 50-253-2552