Learn More
Kingfisher Biotech Inc Canine IL-4 Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0992D100
Encompass
The Canine IL-4 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine IL-4 applications are for cell culture, ELISA standard, and Western Blot Control. The Canine IL-4 yeast-derived recombinant protein can be purchased in multiple sizes. Canine IL-4 Specifications: (Molecular Weight: 12.8 kDa) (Amino Acid Sequence: HNFNITIKEI IKMLNILTAR NDSCMELTVK DVFTAPKNTS DKEIFCRAAT VLRQIYTHNC SNRYLRGLYR NLSSMANKTC SMNEIKKSTL KDFLERLKVI MQKKYYRH (108)) (Gene ID: 403785). For research use only. Made in the USA
Catalog No. 50-241-3528