Learn More
Meso Scale Discovery Canine IL10 UltraSensitive Kit
SDP
Catalog No. 5000078231
Encompass
The Meso Scale Discovery Canine IL-10 Kit is a highly sensitive assay for detecting canine interleukin 10 (IL-10) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide accurate and reproducible quantitation of canine IL-10 with a lower limit of detection of 1 3 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and assay diluent The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78231 Supplier Meso Scale Discovery Supplier No. K154AOC2
The Meso Scale Discovery Canine IL-10 Kit is a highly sensitive assay for detecting canine interleukin 10 (IL-10) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide accurate and reproducible quantitation of canine IL-10 with a lower limit of detection of 1 3 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrators and assay diluent The assay takes approximately 3 5 hours to complete