Learn More
Sino Biological Recombinant Anti-IL-2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 70014R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Canine IL-2 (Catalog#70014-DNAE; Q29416; Ala21-Thr155,147Cys/Ser).
Catalog No. 50-241-0735