Learn More
Sino Biological Anti-IL-5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 70019RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Canine IL-5 / IL5 / Interleukin 5 ( Catalog#70019-D08B; NP_001006951.1; Met1-Ser134). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9123