Learn More
Abcam Canine IL8 ELISA Kit (Interleukin-8), 1 x 96 Tests
Catalog No. 502582903
Encompass_Preferred
Canine IL8 ELISA Kit (Interleukin-8) is a Sandwich (quantitative) ELISA kit for the measurement of Canine IL8 (Interleukin-8) in Dog in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-2903 Supplier Abcam Supplier No. AB1554651X96TESTS
Canine IL8 ELISA Kit (Interleukin-8) is a Sandwich (quantitative) ELISA kit for the measurement of Canine IL8 (Interleukin-8) in Dog in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement