Learn More
American Research Products Inc Canine kidney injury molecule
Catalog No. 5000329956
Encompass
Canine kidney injury molecule 1 (Kim-1) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29956 Supplier American Research Products Inc Supplier No. CSBE13035C
Canine kidney injury molecule 1 (Kim-1) ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich