Learn More
Kingfisher Biotech Inc Canine LIF Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1010D005
Encompass
The Canine Leukemia Inhibitory Factor yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine Leukemia Inhibitory Factor applications are for cell culture, ELISA standard, and Western Blot Control. Canine Leukemia Inhibitory Factor yeast-derived recombinant protein can be purchased in multiple sizes. Canine Leukemia Inhibitory Factor Specifications: (Molecular Weight: 19.7 kDa) (Amino Acid Sequence: SPLPITPVNA TCATRHPCHS NLMNQIRNQL AQLNGSANAL FILYYTAQGE PFPNNLDKLC GPNVTDFPPF HANSTEKTRL VELYRIIAYL GASLGNITRD QKVLNPNALS LHSKLNATAD IMRGLLSNVL CRLCSKYHVA HVDVAYGPDT SGKDVFQKKK LGCQLLGKYK QVIAVVAQAF (180)) (Gene ID: 403449). For research use only. Made in the USA
Catalog No. 50-241-3468