Learn More
Kingfisher Biotech Inc Canine M-CSF Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1720D100
Encompass
The Canine M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Canine M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Canine M-CSF Specifications: (Molecular Weight: 18.5 kDa) (Amino Acid Sequence: EEVSEHCSHM IGNGHLQFLQ QLIDSQMETS CQIAFEFVDQ ELLKDPVCYL KKAFVLMQDI MEDTMRFKDN TPNANVIVKL QELSLRLKSC FTKDYEEQDK ACVRTFYETP LQLLEKIKNV FNETKNLLKK DWNIFSKNCN NSFAKCSSQG HEKQQEGP (158)) (Gene ID: 611795). For research use only. Made in the USA
Catalog No. 50-253-2880