Learn More
American Research Products Inc Canine MatriX metalloproteinas
Catalog No. 5000328479
Encompass
Canine MatriX metalloproteinase 1 MMP-1 ELISA kit from CUSABIO Detection Range 0 18 ng/mL-12 ng/mL Sensitivity 0 04 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28479 Supplier American Research Products Inc Supplier No. CSBE14057C
Canine MatriX metalloproteinase 1 MMP-1 ELISA kit from CUSABIO Detection Range 0 18 ng/mL-12 ng/mL Sensitivity 0 04 ng/mL Sample serum plasma tissue homogenates Method Sandwich