Learn More
American Research Products Inc Canine MatriX metalloproteinas
Catalog No. 5000328691
Encompass
Canine MatriX metalloproteinase 2/Gelatinase A MMP-2 ELISA kit from CUSABIO Detection Range 4 69 ng/mL-300 ng/mL Sensitivity 1 17 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28691 Supplier American Research Products Inc Supplier No. CSBE14056C
Canine MatriX metalloproteinase 2/Gelatinase A MMP-2 ELISA kit from CUSABIO Detection Range 4 69 ng/mL-300 ng/mL Sensitivity 1 17 ng/mL Sample serum plasma tissue homogenates Method Sandwich