Learn More
Raybiotech Inc Canine MCP-1 (CCL2) ELISA Kit
Supplier:  Raybiotech Inc ELCMCP11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 9 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects canine MCP 1. Less than 0.05% cross reactivity with recombinant mouse MCP-1 and recombinant human MCP-1 is observed in this antibody pair.
Catalog No. 50-194-8894