Learn More
Vector Laboratories CANINE MELANOMA CELL TLM-1
SDP
Catalog No. NC3252273
Encompass
TLM-1 is a canine melanoma cell line derived from an oral tumor of a 12-year old male Gordon setter. -Grows in vitro and forms heterotopic tumor xenografts in immunodeficient mice.-Extensively characterized and published onMalignant melanoma is a type of cancer in dogs that affects pigmented-producing cells known as melanocytes. Most malignant melanomas are diagnosed in the skin and mucocutaneous junctions. They tend to grow extremely fast and are likely to spread to other organs including the lungs and liver.
Catalog No. NC3252273 Supplier Vector Laboratories Supplier No. EMN005FP
TLM-1 is a canine melanoma cell line derived from an oral tumor of a 12-year old male Gordon setter. -Grows in vitro and forms heterotopic tumor xenografts in immunodeficient mice.-Extensively characterized and published onMalignant melanoma is a type of cancer in dogs that affects pigmented-producing cells known as melanocytes. Most malignant melanomas are diagnosed in the skin and mucocutaneous junctions. They tend to grow extremely fast and are likely to spread to other organs including the lungs and liver.