Learn More
Sino Biological Recombinant Anti-c-MET/HGFR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 70008R345100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Canine HGFR / c-MET (Catalog#70008-D08H; NP_001002963.1; Met1-Leu935).
Catalog No. 50-240-9092