Learn More
Sino Biological Anti-c-MET/HGFR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 70008RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Canine c-Met / HGFR (Catalog#70008-D08H; NP_001002963.1; Met1-Leu935). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8472