Learn More
Raybiotech Inc Canine MMP-8 ELISA Kit
Supplier:  Raybiotech Inc ELCMMP81
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.07 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Canine Matrix Metallopeptidase 8 (MMP8)
Catalog No. 50-194-8895