Learn More
Sino Biological Recombinant Canine TrkA Protein (His Tag)
Catalog No. 502992831
Encompass_Preferred
A DNA sequence encoding the Canine NTRK1 (A0A8I3P078) (Pro194-Lys410) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-299-2831 Supplier Sino Biological Supplier No. 70101D07H1100
A DNA sequence encoding the Canine NTRK1 (A0A8I3P078) (Pro194-Lys410) was expressed with a polyhistidine tag at the N-terminus.