Learn More
Kingfisher Biotech Inc Canine Oncostatin M Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1183D100
Encompass
The Canine Oncostatin M (OSM) yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine Oncostatin M (OSM) applications are for cell culture, ELISA standard, and Western Blot Control. Canine Oncostatin M (OSM) yeast-derived recombinant protein can be purchased in multiple sizes. Canine Oncostatin M (OSM) Specifications: (Molecular Weight: 20.6 kDa) (Amino Acid Sequence: SCSDKYPELL GQLQKQADFM QHTNTLLDLY IRSQGLDKNG LKEHCRERPG AFPSKDALQR LSRRVFLRTL DTTLGQVLLR LAALEQDIPK AQDLEMLSGV KLNIRGFKNN IHCMAQLLPG SSETTEPTPT SPGASPSPTP TLDTFQRRLE GCRFLHGYHR FMRSVGQVFR EWGKSLSRSR R (181)) (Gene ID: 611921). For research use only. Made in the USA
Catalog No. 50-253-3574