Learn More
American Research Products Inc Canine Parathyroid hormone PT
Catalog No. 5000328152
Encompass
Canine Parathyroid hormone (PTH) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28152 Supplier American Research Products Inc Supplier No. CSBE17575C
Canine Parathyroid hormone (PTH) ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich