Learn More
Bioss Canine Parvovirus
SDP
Supplier:  Bioss BSM72039M
Encompass_Preferred
Canine Parvovirus Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65813