Learn More
ACROBiosystems ACROBiosystems Canine PD-1 / PDCD1 Protein, His Tag (MALS verified)
Supplier:  ACROBiosystems PD1C52H91MG
Encompass_Preferred
Recombinant Protein;1MG;Canine PD-1, His Tag (PD1-C52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gly 168 (Accession # NP_001301026.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.0 kDa. The protein migrates as 30-40 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-210-6212