Learn More
ACROBiosystems ACROBiosystems Canine PD-L1 / B7-H1 Protein, Fc Tag
Catalog No. 502107377
Encompass_Preferred
Recombinant Protein;100UG;Canine PD-L1, Fc Tag (PD1-C52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - His 238 (Accession # E2RKZ5-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7377 Supplier ACROBiosystems Supplier No. PD1C52H3100UG
Recombinant Protein;100UG;Canine PD-L1, Fc Tag (PD1-C52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - His 238 (Accession # E2RKZ5-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.4 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.