Learn More
Sino Biological Anti-PD-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 70109T20100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Canine PD1/PDCD1/CD279 ( Catalog#70109-D08H; XP_022265285.1; Met1-Leu169). PD1/PDCD1/CD279 specific IgG was purified by Canine PD1/PDCD1/CD279 affinity chromatography.
Catalog No. 50-241-1963