Learn More
Sino Biological Recombinant Canine PD-1 Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 70109D27HB100
Encompass_Preferred
A DNA sequence encoding the canine PDCD1 (NP_001301026.1) (Met1-Gly168) was expressed with a C-terminal Avi tag followed by a His tag. The purified protein was biotinylated in vitro.
Catalog No. 50-255-2896