Learn More
Raybiotech Inc Canine PDGF-BB ELISA Kit
Supplier:  Raybiotech Inc ELCPDGFB1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects canine PDGF-BB, it also can recognize human PDGF-BB.
Catalog No. 50-194-8896