Learn More
American Research Products Inc Canine Procalcitonin PCT ELI
Catalog No. 5000328508
Encompass
Canine Procalcitonin (PCT) ELISA Kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 72 pg/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-28508 Supplier American Research Products Inc Supplier No. CSBE17400C
Canine Procalcitonin (PCT) ELISA Kit from CUSABIO Detection Range 46 88 pg/mL-3000 pg/mL Sensitivity 11 72 pg/mL Sample serum plasma cell culture supernates Method Sandwich