Learn More
American Research Products Inc Canine PTH ELISA Kit
Catalog No. 5000311582
Encompass
This Canine Parathyroid Hormone (PTH) ELISA Kit is a precoated Competitive Inhibition ELISA assay designed to detect and quantify the target analyte in serum plasma and other biological fluids This product is for Research Use Only
Catalog No. 50-003-11582 Supplier American Research Products Inc Supplier No. RDPTHC
This Canine Parathyroid Hormone (PTH) ELISA Kit is a precoated Competitive Inhibition ELISA assay designed to detect and quantify the target analyte in serum plasma and other biological fluids This product is for Research Use Only