Learn More
Raybiotech Inc Canine Resistin ELISA Kit
Supplier:  Raybiotech Inc ELCRESISTIN1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 16 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Canine Resistin (RETN)
Catalog No. 50-194-8898