Learn More
Kingfisher Biotech Inc Mouse TNF alpha ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1565D004
Encompass_Preferred
The Mouse TNF alpha ELISA contains capture antibody, standard, and detection antibody for development of a Mouse TNF alpha ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include TNF, DIF, TNF-alpha, TNFA, TNFSF2, Tumor necrosis factor, tumor necrosis factor, TNLG1F, Tumor necrosis factor alpha. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5524