Learn More
Kingfisher Biotech Inc Canine, Equine, Feline, Ferret, and Swine TGF-alpha Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1633005
Encompass
The Canine, Equine, Feline, Ferret, and Swine TGF-alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine, Equine, Feline, Ferret, and Swine TGF-alpha applications are for cell culture, ELISA standard, and Western Blot Control. Canine, Equine, Feline, Ferret, and Swine TGF-alpha yeast-derived recombinant protein can be purchased in multiple sizes. Canine, Equine, Feline, Ferret, and Swine TGF-alpha Specifications: (Molecular Weight: 5.5 kDa) (Amino Acid Sequence: VVSHFNDCPD SHSQFCFHGT CRFLVQEDKP ACVCHSGYVG ARCEHADLLA (50)) (Gene ID: n/a). For research use only. Made in the USA
Catalog No. 50-253-3033