Learn More
Sino Biological Canine TIM-1/HAVCR1 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785805
Encompass_Preferred
The Canine KIM-1 / TIM1 / HAVCR1 ELISA Pair Set is for the quantitative determination of Canine KIM-1 / TIM1 / HAVCR1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Canine KIM-1 / TIM1 / HAVCR1 coated on a 96-well plate. Standards and samples are added to the wells, and any Canine KIM-1 / TIM1 / HAVCR1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Canine KIM-1 / TIM1 / HAVCR1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Canine KIM-1 / TIM1 / HAVCR1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85805 Supplier Sino Biological Supplier No. SEK700015
The Canine KIM-1 / TIM1 / HAVCR1 ELISA Pair Set is for the quantitative determination of Canine KIM-1 / TIM1 / HAVCR1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Canine KIM-1 / TIM1 / HAVCR1 coated on a 96-well plate. Standards and samples are added to the wells, and any Canine KIM-1 / TIM1 / HAVCR1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Canine KIM-1 / TIM1 / HAVCR1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Canine KIM-1 / TIM1 / HAVCR1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.