Learn More
Raybiotech Inc Canine TIMP-2 ELISA Kit
Supplier:  Raybiotech Inc ELCTIMP21
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 20 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Canine Metalloproteinase inhibitor 2.
Catalog No. 50-194-8900