Learn More
Kingfisher Biotech Inc Canine TNF alpha Polyclonal Antibody (100 ug)
SDP
Supplier:  Kingfisher Biotech Inc PB0389D100
Encompass
The Canine TNF alpha polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Biotech Canine TNF alpha ELISA. The Canine TNF alpha polyclonal antibody was produced in goats and is antigen-affinity purified. The reactivity by species is: (Bovine TNFalpha - None) (Canine TNFalpha - Strong) (Caprine TNFalpha - None) (Cynomolgus Monkey TNFalpha - Strong) (Dolphin TNFalpha - None) (Equine TNFalpha - Weak) (Feline TNFalpha - Moderate) (Guinea Pig TNFalpha - None) (Human TNFalpha - Strong) (Mouse TNFalpha - None) (Ovine TNFalpha - None) (Rabbit TNFalpha - Moderate) (Swine TNFalpha - None). For research use only. Made in the USA
Catalog No. 50-233-2405