Learn More
Kingfisher Biotech Inc Canine TNF alpha Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0261D100
Encompass
The Canine TNF alpha yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine TNF alpha applications are for cell culture, ELISA standard, and Western Blot Control. The Canine TNF alpha yeast-derived recombinant protein can be purchased in multiple sizes. Canine TNF alpha Specifications: (Molecular Weight: 17.3 kDa) (Amino Acid Sequence: VKSSSRTPSD KPVAHVVANP EAEGQLQWLS RRANALLANG VELTDNQLIV PSDGLYLIYS QVLFKGQGCP STHVLLTHTI SRFAVSYQTK VNLLSAIKSP CQRETPEGTE AKPWYEPIYL GGVFQLEKGD RLSAEINLPN YLDFAESGQV YFGIIAL (157)) (Gene ID: 403922). For research use only. Made in the USA
Catalog No. 50-241-3792