Learn More
Raybiotech Inc Canine TNF alpha ELISA Kit
Supplier:  Raybiotech Inc ELCTNFA1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects canine TNF-alpha. Other species not determined yet.
Catalog No. 50-194-8901