Learn More
ACROBiosystems ACROBiosystems Canine TNF-alpha Protein, His Tag, active trimer (MALS verified)
Catalog No. 502107370
Encompass_Preferred
Recombinant Protein;1MG;Canine TNF-alpha, His Tag (TNA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P51742-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 19-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Humira at 5 μg/mL (100 μL/well) can bind Canine TNF-alpha, His Tag (Cat. No. TNA-C52H3) with a linear range of 0.3-1 ng/mL (QC tested).;Immobilized Human TNFR1, Fc Tag (Cat. No. TN1-H5251) at 5 μg/mL (100 μL/well) can bind Canine TNF-alpha, His Tag (Cat. No. TNA-C52H3) with a linear range of 2-10 ng/mL (Routinely tested).
Catalog No. 50-210-7370 Supplier ACROBiosystems Supplier No. TNAC52H31MG
Recombinant Protein;1MG;Canine TNF-alpha, His Tag (TNA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P51742-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.6 kDa. The protein migrates as 19-22 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Humira at 5 μg/mL (100 μL/well) can bind Canine TNF-alpha, His Tag (Cat. No. TNA-C52H3) with a linear range of 0.3-1 ng/mL (QC tested).;Immobilized Human TNFR1, Fc Tag (Cat. No. TN1-H5251) at 5 μg/mL (100 μL/well) can bind Canine TNF-alpha, His Tag (Cat. No. TNA-C52H3) with a linear range of 2-10 ng/mL (Routinely tested).