Learn More
Sino Biological Anti-TNF-alpha Antibody, Mouse Monoclonal
Supplier:  Sino Biological 70003MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Canine TNF-alpha (rCanine TNF-alpha; Catalog #70003-DNAE; NP_001003244.4; Val 77-Leu 233). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2529