Learn More
Sino Biological Recombinant Anti-TNF-alpha Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 70003R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Canine TNF-alpha (rCanine TNF-alpha; Catalog#70003-DNAE; NP_001003244.4; Val 77-Leu 233).
Catalog No. 50-240-9129