Learn More
Kingfisher Biotech Inc Canine TNFSF11 (RANKL) Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1106D025
Encompass
The Canine TNFSF11 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine TNFSF11 applications are for cell culture, ELISA standard, and Western Blot Control. Canine TNFSF11 yeast-derived recombinant protein can be purchased in multiple sizes. Canine TNFSF11 Specifications: (Molecular Weight: 19.8 kDa) (Amino Acid Sequence: EKAMMEGSWL EMARRGKTHT QPFAHLTINA TDIPSGSHKV SLSSWYHDRG WAKISNMTFS NGKLIVNQDG FYFLYANICF RHHETSGDLA TEYLQLMVYV TKTSIKIPSS HTLMKGGSTK YWSGNSEFHF YSINVGGFFK LRSGEEISIE VSNPSLLDPD QDATYFGAFK VLDID (175)) (Gene ID: 609418). For research use only. Made in the USA
Catalog No. 50-253-3660