Learn More
American Research Products Inc Canine Tri iodothyronine T3 E
Catalog No. 5000329585
Encompass
Canine Tri-iodothyronine T3 ELISA Kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29585 Supplier American Research Products Inc Supplier No. CSBE07287C
Canine Tri-iodothyronine T3 ELISA Kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma Method Competitive