Learn More
ACROBiosystems ACROBiosystems Canine TROP-2 / TACSTD2 Protein, His Tag
Catalog No. 502107138
Encompass_Preferred
Recombinant Protein;100UG;Canine TROP-2, His Tag (TR2-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 39 - Thr 281 (Accession # XP_852842.5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.4 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7138 Supplier ACROBiosystems Supplier No. TR2C52H4100UG
Recombinant Protein;100UG;Canine TROP-2, His Tag (TR2-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 39 - Thr 281 (Accession # XP_852842.5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 29.4 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.