Learn More
Med Vet International Canine "V" Notch Elevator, CVD, Inside Grooved, J0899D, Each
Catalog No. 5000584076
Encompass_Preferred
Modified with a "V" cut into the end tip of this instrument, this helps the tip stay on the tooth surface and guides into the socket, preventing it from slipping and damaging the gingiva. The curved tip is designed to follow the root curves of teeth, (i.e., canine and incisor teeth). The elevator tip can be on either the outside or inside of the curved end.
Catalog No. 50-005-84076 Supplier Med Vet International Supplier No. JOR899D
Modified with a "V" cut into the end tip of this instrument, this helps the tip stay on the tooth surface and guides into the socket, preventing it from slipping and damaging the gingiva. The curved tip is designed to follow the root curves of teeth, (i.e., canine and incisor teeth). The elevator tip can be on either the outside or inside of the curved end.