Learn More
Kingfisher Biotech Inc Canine VEGF-A Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0252D025
Encompass
The Canine VEGF-A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine VEGF-A applications are for cell culture, ELISA standard, and Western Blot Control. The Canine VEGF-A yeast-derived recombinant protein can be purchased in multiple sizes. Canine SCF Specifications: (Molecular Weight: 22.0 kDa) (Amino Acid Sequence: APMAGGEHKPHEVVKFMDVYQRSYCRPIETLVDIFQEYPDEIEYIFKPSCVPLMRCGGCCNDEGLECVPTEEFNITMQIMRIKPHQGQHIGEMSFLQHSKCECRPKKDRARQEKKSIRGKGKGQKRKRKKSRYKPWSVPCGPCSERRKHLFVQDPQTCKCSCKNTDSRCKARQLELNERTCRCDKPRR (188)) (Gene ID: 403802). For research use only. Made in the USA
Catalog No. 50-241-3214