Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 55107MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: DG1/447. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Gastrointestinal Stromal Tumor (GIST) or testicular germ cell tumor. Melanocytes in the basal layer of the epidermis, mast cells in the dermis of normal skin. Cellular Staining: Cell Surface, Cytoplasm. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human DOG-1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 114kDa.Chromosome: 11q13.3. Gene Symbol: TMEM16A. Gene ID: 55107. Ensemble: ENSG00000131620. Uniprot: NP_011543428.
Catalog No. 50-188-5074