Learn More
Med Vet International NobIVac Canine Vaccine 1-Dappv, Subcutaneous/intramuscular Injection, 25X1 Dose Tray
Catalog No. 502232559
Encompass_Preferred
Recommended for use in healthy dogs as an aid in the prevention of disease caused by canine distemper virus, adenovirus type 1 (hepatitis) and adenovirus type 2 (respiratory disease), canine parainfluenza virus, and canine parvovirus. Proven 1-year vaccine that offers broad protection against canine parvovirus (CPV), adenovirus type 1 (hepatitis) and type 2 (respiratory disease), parainfluenza, and distemper. Recommended for use in healthy dogs 6 weeks of age or older. Administration: Subcutaneous or intramuscular injection. Available in a 25 x 1 mL dose. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. *Next Day Air Available With Additional Charges*
Catalog No. 50-223-2559 Supplier Med Vet International Supplier No. PHV1DAPPV
Recommended for use in healthy dogs as an aid in the prevention of disease caused by canine distemper virus, adenovirus type 1 (hepatitis) and adenovirus type 2 (respiratory disease), canine parainfluenza virus, and canine parvovirus. Proven 1-year vaccine that offers broad protection against canine parvovirus (CPV), adenovirus type 1 (hepatitis) and type 2 (respiratory disease), parainfluenza, and distemper. Recommended for use in healthy dogs 6 weeks of age or older. Administration: Subcutaneous or intramuscular injection. Available in a 25 x 1 mL dose. Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. *Next Day Air Available With Additional Charges*