Learn More
Med Vet International Nobivac, Merck, Canine Vaccine 1-DPPvL2, 25x1 Dose Tray
Catalog No. 502188577
Encompass_Preferred
**only offer 6+ month dating** Protects against all known strains of CPV, including CPV-2c High antigenic mass (titer), low passage parvovirus vaccine Canine vaccine containing CPV-2b, one of the most prevalent field strains of parvovirus Protects against two types of adenovirus that cause hepatitis and respiratory disease in dogs Provides disease coverage for commonly spread canine viruses, distemper, adenovirus types 1 and 2, parainfluenza and parvovirus in 1 formula Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. *Next Day Air Available With Additional Charges*
Catalog No. 50-218-8577 Supplier Med Vet International Supplier No. PHV1DAPPVL2
**only offer 6+ month dating** Protects against all known strains of CPV, including CPV-2c High antigenic mass (titer), low passage parvovirus vaccine Canine vaccine containing CPV-2b, one of the most prevalent field strains of parvovirus Protects against two types of adenovirus that cause hepatitis and respiratory disease in dogs Provides disease coverage for commonly spread canine viruses, distemper, adenovirus types 1 and 2, parainfluenza and parvovirus in 1 formula Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. *Next Day Air Available With Additional Charges*