Learn More
Med Vet International Nobivac Intra-Trac 3 Canine Intranasal Vaccine, 25x1 Dose Tray
Catalog No. 502188613
Encompass_Preferred
Kennel Cough Vaccine for Dogs & Puppies Combination vaccine for dogs For healthy dogs and puppies 3 weeks of age or older Protects against canine adenovirus type 2, parainfluenza, and Bordetella bronchiseptica infection 3-way protection against Kennel Cough Modified live virus intranasal vaccine, requires mixing Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. Next day air available for additional charge.
Catalog No. 50-218-8613 Supplier Med Vet International Supplier No. PHVINTRATRAC3
Kennel Cough Vaccine for Dogs & Puppies Combination vaccine for dogs For healthy dogs and puppies 3 weeks of age or older Protects against canine adenovirus type 2, parainfluenza, and Bordetella bronchiseptica infection 3-way protection against Kennel Cough Modified live virus intranasal vaccine, requires mixing Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. Next day air available for additional charge.