Learn More
Braintree Scientific Braintree Sci Anesthesia Gas Filter, each
SDP
Catalog No. NC9563652
Encompass
Braintree Sci Anesthesia Gas Filter, a sensible answer to anesthesia gas problems in the operating room, the charcoal anesthesia gas filter was specifically designed to remove waste anesthesia gases from the operating room environment. Economical to use, with an average useful life of 12 to 15 hours. Canister isinterchangeable with sytems that use F/Air canisters.BSci-Airwillremove no less than 50 grams of pure halogenated anesthetic gases and can then be conveniently discarded. **Sold for research purposes only and not for human use**
Catalog No. NC9563652 Supplier Braintree Scientific Supplier No. BSCIAIREA
Braintree Sci Anesthesia Gas Filter, a sensible answer to anesthesia gas problems in the operating room, the charcoal anesthesia gas filter was specifically designed to remove waste anesthesia gases from the operating room environment. Economical to use, with an average useful life of 12 to 15 hours. Canister isinterchangeable with sytems that use F/Air canisters.BSci-Airwillremove no less than 50 grams of pure halogenated anesthetic gases and can then be conveniently discarded. **Sold for research purposes only and not for human use**