Learn More
Custom Biogenic Systems Custom Biogenic Systems ZC022 Canister, Aluminum, 500 mL Capacity
Catalog No. NC0792036
Encompass
The Custom Biogenic Systems aluminum canister is designed for effective cryogenic storage, ensuring durability for repeated use in ultra-low temperatures. It supports organized storage of biological samples, particularly in Baxter Fenwal containers. This robust canister's compatibility and construction differentiate it from standard alternatives.
Catalog No. NC0792036 Supplier Custom Biogenic Systems Supplier No. ZC022
The Custom Biogenic Systems aluminum canister is designed for effective cryogenic storage, ensuring durability for repeated use in ultra-low temperatures. It supports organized storage of biological samples, particularly in Baxter Fenwal containers. This robust canister's compatibility and construction differentiate it from standard alternatives.